Learn More
S100A3 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98279MFV500
This item is not returnable.
View return policy
Description
S100A3 Polyclonal antibody specifically detects S100A3 in Mouse samples. It is validated for Western Blot, ELISA
Specifications
| S100A3 | |
| mFluor Violet 500 SE | |
| Protein S-100E, S100 calcium binding protein A3, S100 calcium-binding protein A3S100Eprotein S100-A3 | |
| Store at 4C in the dark. |
| ELISA | |
| 50mM Sodium Borate | |
| 6274 | |
| Purified |