missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A4 Antibody (S100A4/1482), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54400APCCY7
This item is not returnable.
View return policy
Description
S100A4 Monoclonal specifically detects S100A4 in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifications
| S100A4 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| S100A4 | |
| Recombinant fragment of human S100A4 protein (aa1-200) (exact sequence is proprietary) (Uniprot: P26447) | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 κ |
| Flow Cytometry | |
| S100A4/1482 | |
| Flow Cytometry, Flow (Intracellular) | |
| 18A2, Calvasculin, CAPLS100 calcium binding protein A4 (calcium protein, calvasculin, metastasin, fibroblast-specific protein-1,42A, FSP1, leukemia multidrug resistance associated protein, malignant transformation suppression 1, Metastasin, MTS1, murine placental homolog), P9KA, PEL98, Placental calcium-binding protein, Protein Mts1, protein S100-A4, S100 calcium binding protein A4, S100 calcium-binding protein A4, S100 calcium-binding protein A4 (calcium protein, calvasculin, metastasin, murine placental homolog) | |
| Mouse | |
| Protein A or G purified | |
| Cancer | |
| 6275.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction