Learn More
S100A4 Mouse anti-Human, CoraLite 594, Clone: 2G11B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66489-100UL
Description
The S100A4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human S100A4. This antibody recognizes human, mouse antigen. The S100A4 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.
Specifications
| S100A4 | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 6275 | |
| IgG2a |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 2G11B4 | |
| Mouse | |
| Primary | |
| Human, Mouse |