Learn More
S100A4 Mouse anti-Human, CoraLite 594, Clone: 2G11B4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-66489-100UL
Product Code. 16821046
  • € 450.00 / 100µL
Description
The S100A4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human S100A4. This antibody recognizes human, mouse antigen. The S100A4 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.
Specifications
Specifications
S100A4
Monoclonal
CoraLite 594
100 μL
6275
IgG2a
Immunofluorescence, Flow Cytometry (Intracellular Staining)
2G11B4