missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93842-0.1ml
This item is not returnable.
View return policy
Description
S100A5 Polyclonal antibody specifically detects S100A5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| S100A5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:100-1:200, Immunohistochemistry-Paraffin | |
| Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-92 of human S100A5 (NP_002953.2). METPLEKALTTMVTTFHKYSGREGSKLTLSRKELKELIKKELCLGEMKESSIDDLMKSLDKNSDQEIDFKEYSVFLTMLCMAYNDFFLEDNK | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 6276 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction