missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A5 Antibody (OTI4C9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45660
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
S100A5 Monoclonal specifically detects S100A5 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| S100A5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5 | |
| Mouse | |
| 12.6 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI4C9 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| S100A5 | |
| Full length human recombinant protein of human S100A5 (NP_002953) produced in 293T cell. | |
| Immunogen affinity purified | |
| RUO | |
| 6276 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction