missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A5 Antibody (S100A5/7472), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23994UV
This item is not returnable.
View return policy
Description
S100A5 Monoclonal antibody specifically detects S100A5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| S100A5 | |
| Monoclonal | |
| DyLight 350 | |
| Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5 | |
| Recombinant fragment (around aa1-92) of human S100A5 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 6276 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| S100A5/7472 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction