missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A5 Antibody (S100A5/7475), Novus Biologicals™
Mouse Monoclonal Antibody
€ 232.00 - € 462.00
Specifications
| Antigen | S100A5 |
|---|---|
| Clone | S100A5/7475 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630535
|
Novus Biologicals
NBP3-23848-20ug |
20 μg |
€ 232.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18664015
|
Novus Biologicals
NBP3-23848-100ug |
100 μg |
€ 462.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S100A5 Monoclonal antibody specifically detects S100A5 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| S100A5 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Neuroscience, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 6276 | |
| IgG2a κ | |
| Protein A or G purified |
| S100A5/7475 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5 | |
| Recombinant fragment (around aa1-92) of human S100A5 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title