missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17924-1-AP-150UL
This item is not returnable.
View return policy
Description
Binds calcium, zinc and copper. One subunit can simultaneously bind 2 calcium ions or 2 copper ions plus 1 zinc ion. Calcium and copper ions compete for the same binding sites. Engelkamp D., Proc. Natl. Acad. Sci. U.S.A. 90:6547-6551(1993). Gregory S.G., Nature 441:315-321(2006).Specifications
| S100A5 | |
| Polyclonal | |
| Unconjugated | |
| S100A5 | |
| Protein S 100D, Protein S100 A5, S100A5, S100D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20199, 295211, 6276 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P33763, P63083, P63084 | |
| S100A5 | |
| S100A5 Fusion Protein Ag12342 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction