missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100A6 Recombinant Rabbit Monoclonal Antibody (0J1Q5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538070
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, C2C12, HUVEC, Mouse lung, Mouse spleen, Rat lung, Rat spleen Immunogen sequence: MACPLDQAIG LLVAIFHKYS GREGDKHTLS KKELKELIQK ELTIGSKLQD AEIARLMEDL DRNKDQEVNF QEYVTFLGAL ALIYNEALKG.
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in stimulation of Ca2+-dependent insulin release, stimulation of prolactin secretion, and exocytosis. Chromosomal rearrangements and altered expression of this gene have been implicated in melanoma.
Specifications
| S100A6 | |
| Recombinant Monoclonal | |
| 0.23 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05964, P06703, P14069 | |
| S100A6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human S100A6 (P06703). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0J1Q5 | |
| Unconjugated | |
| S100A6 | |
| 2A9; 5B10; CABP; Cacy; calcium binding protein A6 (calcyclin); calcyclin; Growth factor-inducible protein 2A9; lung 10 kDa protein; MLN 4; PRA; Prolactin receptor-associated protein; protein S100-A6; S100 calcium binding protein A6; S100 calcium binding protein A6 (calcyclin); S100 calcium-binding protein A6; S100 calcium-binding protein A6 (calcyclin); S100a6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20200, 6277, 85247 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction