Learn More
S100A7/Psoriasin Antibody (47C1068), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-24911AF700
Product Code. 18109623
  • € 659.00 / 0.10mL
Description
S100A7/Psoriasin Monoclonal specifically detects S100A7/Psoriasin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S100A7/Psoriasin
Monoclonal
Alexa Fluor 700
HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c
Mouse
Protein G purified
RUO
Primary
In confluent MCF10A cells, an 11 kDa band is observed.
Store at 4C in the dark.
IgG1 κ
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
47C1068
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
S100A7
This antibody was developed against recombinant psoriasin/HID-5 protein.
0.1 mL
Cancer
6278