missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A7/Psoriasin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99720-100ul
This item is not returnable.
View return policy
Description
S100A7/Psoriasin Polyclonal antibody specifically detects S100A7/Psoriasin in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
Specifications
| S100A7/Psoriasin | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c | |
| Produced in rabbits immunized with purified, recombinant Human S100A7/Psoriasin (Accession#: NP_002954.2; Met 1-Gln 101) | |
| 100 μg | |
| Cancer | |
| 6278 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction