missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A8 Antibody (101), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89805MFV610
This item is not returnable.
View return policy
Description
S100A8 Monoclonal antibody specifically detects S100A8 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| S100A8 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 101 | |
| Immunohistochemistry-Paraffin | |
| 60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A8 (Accession#: NP_002955.2; Met 1-Glu 93). | |
| 0.1 mL | |
| Cancer | |
| 6279 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction