missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A8/S100A9 Heterodimer Antibody (900027), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB45702U
This item is not returnable.
View return policy
Description
S100A8/S100A9 Heterodimer Monoclonal antibody specifically detects S100A8/S100A9 Heterodimer in Human samples. It is validated for ELISA, Immunohistochemistry, Flow Cytometry, CyTOF
Specifications
| S100A8/S100A9 Heterodimer | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| E. coli-derived recombinant human S100A8/S100A9 heterodimer, Met1-Glu93 (S100A8) and Thr2-Pro114 (S100A9), Accession # P05109 (S100A8) and P06702 (S100A9) | |
| 0.1 mL | |
| Cancer | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Flow Cytometry, CyTOF | |
| 900027 | |
| ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction