missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A8/S100A9 Heterodimer Antibody (900028), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB45703E
This item is not returnable.
View return policy
Description
S100A8/S100A9 Heterodimer Monoclonal antibody specifically detects S100A8/S100A9 Heterodimer in Human samples. It is validated for ELISA, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| S100A8/S100A9 Heterodimer | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| E. coli-derived recombinant human S100A8/S100A9 heterodimer, Met1-Glu93 (S100A8) & Thr2-Pro114 (S100A9), Accession # P05109 (S100A8) and P06702 (S100A9) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
| ELISA, Flow Cytometry (Intracellular Staining), CyTOF | |
| 900028 | |
| ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction