Learn More
S100A9 Antibody (47-8D3 (DF3973)), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47980MFV610
This item is not returnable.
View return policy
Description
S100A9 Monoclonal antibody specifically detects S100A9 in Human, Mouse, Rat samples. It is validated for Flow Cytometry