missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100B Antibody (S1-61), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50282B
This item is not returnable.
View return policy
Description
S100B Monoclonal specifically detects S100B in Human samples. It is validated for Immunohistochemistry.
Specifications
| S100B | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| S100B | |
| S100B protein conjugated to methylated BSA | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Biologically Active Proteins, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Epigenetics, Hypoxia, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction, Stem Cells | |
| 6285 | |
| Store at 4C in the dark. | |
| IgG1 |
| Immunohistochemistry | |
| S1-61 | |
| Immunohistochemistry | |
| beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction