Learn More
S100B Monoclonal Antibody (OTI3G6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA807235S
Encompass_Preferred
 View more versions of this product
Product Code. 17507623
  • € 242.00 / 30µL
Description
S100B Monoclonal antibody specifically detects S100B in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
S100B
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P02749
Mouse
Affinity chromatography
RUO
350
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI3G6
Unconjugated
APOH
AI850290; Bpb; hypothetical protein LOC525716; NEF; protein S100-B; S100; S100 calcium binding protein B; S100 calcium binding protein beta (neural); S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S-100 calcium-binding protein beta subunit; S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S100 protein beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100B; S100-B; S100beta; S100P
Human recombinant protein fragment corresponding to amino acids 120-345 of human APOH produced in E.coli.
30 μL