missing translation for 'onlineSavingsMsg'
Learn More

S100B Mouse anti-Human, Rabbit, Rat, Clone: SH-B1, Abnova™

Product Code. 16108829
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16108829 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16108829 Supplier Abnova Supplier No. MAB12455.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against full length native of bovine S100B.

Specifications

Antigen S100B
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SH-B1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length native of bovine S100B.
Dilution Immunohistochemistry (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from ascites fluid (1.2% sodium acetate, 2mg BSA, 0.01mg sodium azide)
Gene Alias MGC128275
Gene Symbols S100B
Host Species Mouse
Immunogen Native purified S100B from bovine brain.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 525716
Target Species Human, Rabbit, Rat
Content And Storage Store at −20°C. After reconstitution with 1mLof 1.2% sodium acetate or neutral PBS and concentration will be 100μg/mL, store at −20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.