missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100P Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 401.00 - € 672.00
Specifications
| Antigen | S100P |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672024
|
Novus Biologicals
NBP3-21384-25ul |
25 μg |
€ 401.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18622624
|
Novus Biologicals
NBP3-21384-100ul |
100 μg |
€ 672.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S100P Polyclonal antibody specifically detects S100P in Human samples. It is validated for ImmunofluorescenceSpecifications
| S100P | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS, pH 7.2, 40% glycerol | |
| 6286 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| MIG9, migration-inducing gene 9, Protein S100-E, S100 calcium binding protein P, S100 calcium-binding protein Pprotein S100-P, S100E | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: FIVFVAAITSACHKYFEKAGL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel