missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100P Antibody (S100P/4386R), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08769CL1
This item is not returnable.
View return policy
Description
S100P Monoclonal antibody specifically detects S100P in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| S100P | |
| Monoclonal | |
| CoraFluor 1 | |
| MIG9, migration-inducing gene 9, Protein S100-E, S100 calcium binding protein P, S100 calcium-binding protein Pprotein S100-P, S100E | |
| Recombinant fragment corresponding to C-terminal of human S100P protein (Uniprot: P25815) | |
| 0.1 mL | |
| Cancer | |
| 6286 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| S100P/4386R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction