missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100P Antibody (S100P/7375), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20929AF647
This item is not returnable.
View return policy
Description
S100P Monoclonal antibody specifically detects S100P in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| S100P | |
| Monoclonal | |
| Alexa Fluor 647 | |
| MIG9, migration-inducing gene 9, Protein S100-E, S100 calcium binding protein P, S100 calcium-binding protein Pprotein S100-P, S100E | |
| Recombinant fragment (around aa1-95) of human S100P protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer | |
| 6286 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| S100P/7375 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction