missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100P Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11803-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21; however, this gene is located at 4p16. This protein, in addition to binding Ca2+, also binds Zn2+ and Mg2+. This protein may play a role in the etiology of prostate cancer.Specifications
| S100P | |
| Polyclonal | |
| Unconjugated | |
| S100P | |
| MIG9, Protein S100 E, Protein S100 P, S100 calcium binding protein P, S100E, S100P | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6286 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.58 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P25815 | |
| S100P | |
| S100P Fusion Protein Ag2379 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction