missing translation for 'onlineSavingsMsg'
Learn More

S1p receptor agonist 1, MedChemExpress

Product Code. p-200100638
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494556

Brand: Medchem Express HY101265

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress S1p receptor agonist 1 is a potent and orally active S1P receptor agonist, exhibits an activity of inducing S1P1 internalization (EC50=9.83 nM). S1p receptor agonist 1 has the potential for the study of arthritis and EAE (experimental autoimmune encephalitis). S1p receptor agonist 1 is extracted from patent WO2015039587A1, Compound 2.

S1p receptor agonist 1 is a potent and orally active S1P receptor agonist, exhibits an activity of inducing S1P1 internalization (EC50=9.83 nM). S1p receptor agonist 1 has the potential for the study of arthritis and EAE (experimental autoimmune encephalitis). S1p receptor agonist 1 is extracted from patent WO2015039587A1, Compound 2.

Specifications

Chemical Name or Material S1p receptor agonist 1
CAS 1514888-56-2
Color White
Purity Grade Notes Research
Molecular Formula C23H24FN3O3
Quantity 100 mg
Solubility Information DMSO : 6.8 mg/mL (16.61 mM; Need ultrasonic and warming)
SMILES O=C(C1CN(CC2=CC=C(C3=NOC(C4=CC=C(CC(C)C)C=C4)=N3)C=C2F)C1)O
Molecular Weight (g/mol) 409.45
Formula Weight 409.45
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.