missing translation for 'onlineSavingsMsg'
Learn More

S1p receptor agonist 1, MedChemExpress

Product Code. 16469846
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469846 50 mg 50mg
16494556 100 mg 100mg
16483308 10 mg 10mg
16406037 5 mg 5mg
16400749 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16469846 Supplier Medchem Express Supplier No. HY101265

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress S1p receptor agonist 1 is a potent and orally active S1P receptor agonist, exhibits an activity of inducing S1P1 internalization (EC50=9.83 nM). S1p receptor agonist 1 has the potential for the study of arthritis and EAE (experimental autoimmune encephalitis). S1p receptor agonist 1 is extracted from patent WO2015039587A1, Compound 2.

S1p receptor agonist 1 is a potent and orally active S1P receptor agonist, exhibits an activity of inducing S1P1 internalization (EC50=9.83 nM). S1p receptor agonist 1 has the potential for the study of arthritis and EAE (experimental autoimmune encephalitis). S1p receptor agonist 1 is extracted from patent WO2015039587A1, Compound 2.

Specifications

Chemical Name or Material S1p receptor agonist 1
CAS 1514888-56-2
Color White
Purity Grade Notes Research
Molecular Formula C23H24FN3O3
Quantity 50 mg
Solubility Information DMSO : 6.8 mg/mL (16.61 mM; Need ultrasonic and warming)
SMILES O=C(C1CN(CC2=CC=C(C3=NOC(C4=CC=C(CC(C)C)C=C4)=N3)C=C2F)C1)O
Molecular Weight (g/mol) 409.45
Formula Weight 409.45
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.