missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ S1P2/EDG-5/S1PR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-27900
This item is not returnable.
View return policy
Description
S1P2/EDG-5/S1PR2 Polyclonal antibody specifically detects S1P2/EDG-5/S1PR2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| S1P2/EDG-5/S1PR2 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| AGR16, EDG5, EDG-5, endothelial differentiation G-protein coupled receptor 5, endothelial differentiation, sphingolipid G-protein-coupled receptor, 5, Gpcr13, H218, LPB2, S1P receptor 2, S1P receptor Edg-5, S1P receptor EDG5, S1P2, sphingosine 1-phosphate receptor 2, sphingosine 1-phosphate receptor Edg-5, sphingosine-1-phosphate receptor 2 | |
| S1P2/EDG-5/S1PR2 antibody was raised against an 16 amino acid peptide near the carboxy terminus of human S1P2/EDG-5/S1PR2. The immunogen is located within amino acids 270 - 320 of S1P2/EDG-5/S1PR2. | |
| 0.1 mg | |
| GPCR | |
| 9294 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 - 2 μg/mL, ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 10 μg/mL | |
| O95136 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction