missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S1P3/EDG-3 Antibody (776808) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7524JF585
This item is not returnable.
View return policy
Description
S1P3/EDG-3 Monoclonal antibody specifically detects S1P3/EDG-3 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| S1P3/EDG-3 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| EDG-3, EDG3FLJ37523, Endothelial differentiation G-protein coupled receptor 3, endothelial differentiation, sphingolipid G-protein-coupled receptor, 3, FLJ93220, G protein-coupled receptor, endothelial differentiation gene-3, LPB3, MGC71696, S1P receptor 3, S1P receptor Edg-3, S1P receptor EDG3, S1P3, sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3, sphingosine-1-phosphate receptor 3 | |
| Human S1P3/EDG-3 transfected NS0 cells,, Accession # Q99500 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 776808 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, GPCR, Lipid and Metabolism, Signal Transduction | |
| 1903 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction