missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S1P4/EDG-6 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24500AF488
This item is not returnable.
View return policy
Description
S1P4/EDG-6 Polyclonal antibody specifically detects S1P4/EDG-6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| S1P4/EDG-6 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| EDG6S1P receptor Edg-6, Endothelial differentiation G-protein coupled receptor 6, endothelial differentiation, G-protein-coupled receptor 6, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor6,endothelial differentiation, G protein coupled receptor 6, S1P receptor 4, S1P4LPC1, SLP4, sphingosine 1-phosphate receptor 4, Sphingosine 1-phosphate receptor Edg-6, sphingosine-1-phosphate receptor 4 | |
| A portion of amino acids 1-50 of human Sphingolipid Receptor Edg6/S1P4 was used as the immunogen. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, GPCR, Signal Transduction | |
| 8698 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction