missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S1P5/EDG-8 Antibody (282503), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3964Z
This item is not returnable.
View return policy
Description
S1P5/EDG-8 Monoclonal antibody specifically detects S1P5/EDG-8 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| S1P5/EDG-8 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 282503 | |
| Flow Cytometry, CyTOF-ready | |
| Edg-8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, S1P receptor 5, S1P5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2 | |
| BaF3 mouse pro-B cell line transfected with human S1P5/EDG-8, Met1-Asp398, Accession # Q9H228 | |
| 0.1 mL | |
| GPCR | |
| 53637 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction