missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Mouse CD197 (4B12), Proteintech
Rat Monoclonal Antibody
Brand: Proteintech 65158-1-IG-100UG
This item is not returnable.
View return policy
Description
The CD197 antibody from Proteintech is a rat monoclonal antibody generated with mouse ccr7 transfected cell line of mouse CD197. This antibody recognizes mouse antigen. The CD197 antibody has been validated for the following applications: FC analysis.Specifications
| CD197 | |
| Monoclonal | |
| 1 mg/mL | |
| Rat | |
| Purified by protein-G affinity chromatography | |
| RUO | |
| 12775 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| 4B12 | |
| Unconjugated | |
| Mouse CCR7 Transfected Cell Line | |
| 100 μg | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction