missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S1PR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102053
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total EDG5.
This gene encodes a member of the G protein-coupled receptors, as well as the EDG family of proteins. The encoded protein is a receptor for sphingosine 1-phosphate, which participates in cell proliferation, survival, and transcriptional activation. Defects in this gene have been associated with congenital profound deafness.
Specifications
| S1PR2 | |
| Polyclonal | |
| Unconjugated | |
| S1PR2 | |
| agr16; edg5; edg-5; S1PR2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14739, 29415, 9294 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95136, P47752, P52592 | |
| S1PR2 | |
| A synthesized peptide derived from human S1PR2(Accession O95136), corresponding to amino acid residues K269-G319. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction