missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S5a/Angiocidin Antibody (CPTC-PSMD4-3), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08271R
This item is not returnable.
View return policy
Description
S5a/Angiocidin Monoclonal antibody specifically detects S5a/Angiocidin in Human samples. It is validated for Western Blot, Protein Array
Specifications
| S5a/Angiocidin | |
| Monoclonal | |
| DyLight 550 | |
| 26S proteasome non-ATPase regulatory subunit 4;AF;AF-1;ASF;MCB1;PSMD4;pUB-R5;Rpn10;S5A | |
| Recombinant full-length human S5a/Angiocidin protein (Uniprot: P55036) | |
| 100 μg | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 5710 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Protein Array | |
| CPTC-PSMD4-3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction