missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S5a/Angiocidin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-90821-25ul
This item is not returnable.
View return policy
Description
S5a/Angiocidin Polyclonal specifically detects S5a/Angiocidin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Proteasome 19S S5A | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| P55036 | |
| PSMD4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VCHSKTRSNPENNVGLITLANDCEVLTTLTPDTGRILSKLHTVQPKGKITFCTGI | |
| 25 μL | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 5710 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 26S proteasome non-ATPase regulatory subunit 4, 26S proteasome regulatory subunit S5A, AF-1,26S protease subunit S5a, AFMCB1, angiocidin, Antisecretory factor 1, ASF, multiubiquitin chain binding protein, Multiubiquitin chain-binding protein, proteasome (prosome, macropain) 26S subunit, non-ATPase, 4, pUB-R5,26S proteasome regulatory subunit RPN10, Rpn10, RPN10 homolog, S5A, S5a/antisecretory factor protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction