missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SA1 Antibody, FITC, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-299F
This item is not returnable.
View return policy
Description
SA1 Polyclonal antibody specifically detects SA1 in Human samples. It is validated for Immunofluorescence
Specifications
| SA1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| cohesin subunit SA-1, nuclear protein stromal antigen 1, SA1, stromal antigen 1 | |
| A 35 amino acid synthetic peptide, which represented a portion of human Stromal Antigen 1 encoded in part by exons 35 and 36 (LocusLink ID 10274). | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair, Mitotic Regulators | |
| 10274 | |
| Store at 4C in the dark. | |
| IgG |
| Immunofluorescence | |
| FITC | |
| PBS | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction