Learn More
Saccharin N-(2-Acetic Acid Ethyl Ester)(Piroxicam Impurity E), TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate |
| CAS | 24683-20-3 |
| Molecular Formula | C11 H11 N O5 S |
| Quantity | 100 mg |
| Synonym | Ethyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate, Piroxicam Imp. E (EP) |
| InChI Formula | InChI=1S/C11H11NO5S/c1-2-17-10(13)7-12-11(14)8-5-3-4-6-9(8)18(12,15)16/h3-6H,2,7H2,1H3 |
| SMILES | CCOC(=O)CN1C(=O)c2ccccc2S1(=O)=O |
| IUPAC Name | ethyl 2-(1,1,3-trioxo-1,2-benzothiazol-2-yl)acetate |
| Molecular Weight (g/mol) | 269.27 |
| Formula Weight | 269.0358 |