Learn More
Sampling Systems™ Sack Samplers
Shop All Sampling Systems Products
Click to view available options
Length (Metric):
400 mm
420 mm
Tip Type:
End Cap
End with Bottle Adapter
Plain End
Unit Size:
Each
Description
- Made from high quality 316L stainless steel
- Single point
- For taking samples from sacks
Specifications
Specifications
| Product Type | Sack Sampler |
| Material | Stainless Steel (316L) |
| Tip Type | End Cap |
| For Use With (Application) | Sampling |
| Diameter (Metric) | 25 mm |
| Diameter (Metric) Core | 22 mm |
| Length (Metric) | 420 mm |