missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SACM1L Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97655H
This item is not returnable.
View return policy
Description
SACM1L Polyclonal specifically detects SACM1L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SACM1L | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DKFZp686A0231, EC 3.1.3, EC 3.1.3.36, EC 3.1.3.77, KIAA0851suppressor of actin 1, phosphatidylinositide phosphatase SAC1, SAC1 suppressor of actin mutations 1-like (yeast), SAC1EC 3.1.3.-, Suppressor of actin mutations 1-like protein | |
| Produced in rabbits immunized with E. coli-derived Human SACM1L fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 22908 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction