missing translation for 'onlineSavingsMsg'
Learn More

Sacubitril hemicalcium salt, MedChemExpress

Product Code. 16482587
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482587 50 mg 50mg
16496488 10 mg 10mg
16456756 100 mg 100mg
16449077 5 mg 5mg
16421776 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16482587 Supplier Medchem Express Supplier No. HY15407A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sacubitril hemicalcium salt (AHU-377 hemicalcium salt) is a potent NEP inhibitor with an IC50 of 5 nM. Sacubitril hemicalcium salt is a component of the heart failure medicine LCZ696.

Sacubitril hemicalcium salt (AHU-377 hemicalcium salt) is a potent NEP inhibitor with an IC50 of 5 nM. Sacubitril hemicalcium salt is a component of the heart failure medicine LCZ696.

Specifications

Chemical Name or Material Sacubitril hemicalcium salt
Physical Form Solid
CAS 1369773-39-6
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C24H28Ca0.5NO5
For Use With (Application) Neuroscience-Neuromodulation
Synonym AHU-377 hemicalcium salt
Solubility Information DMSO : 125 mg/mL (290.35 mM; Need ultrasonic)
SMILES O=C(CCC([O-])=O)N[C@H](CC1=CC=C(C2=CC=CC=C2)C=C1)C[C@@H](C)C(OCC)=O.[0.5Ca2+]
Molecular Weight (g/mol) 430.52
Formula Weight 430.52
Percent Purity 98.41%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.