missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAE2 Antibody (OTI4A3), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73973AF405
This item is not returnable.
View return policy
Description
SAE2 Monoclonal specifically detects SAE2 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| SAE2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| UBA2 | |
| Full length recombinant protein of human SAE2 (NP_005490) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI4A3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Anthracycline-associated resistance ARX, ARX, EC 6.3.2, EC 6.3.2.-, FLJ13058, HRIHFB2115, SAE2SUMO-1 activating enzyme subunit 2, SUMO1 activating enzyme subunit 2, SUMO-activating enzyme subunit 2, UBA2, ubiquitin-activating enzyme E1 homolog, Ubiquitin-like 1-activating enzyme E1B, ubiquitin-like modifier activating enzyme 2, UBLE1B | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 10054.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction