Learn More
Hirschmann™ Safe-Cone Filters for Labopette™
For the prevention of cross contamination of liquid samples or vapours when pipetting radioactive, biohazardous or corrosive samples.
€ 70.20 - € 70.55
Specifications
Type Safety filter
For Use With (Application) Protection against cross-contamination
View More Specs
Products 4
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10233151
View Documents Promotion Details
Hirschmann™
9470010
Labopette models 9475404, 9475405, 9475511, 9475512, 9475513
€ 70.20
Pack of 50
View alternative products
Save Up to 
»
Estimated Shipment: 22-04-2024
to see stock.
Add to basket
10012223
View Documents Promotion Details
Hirschmann™
9470013
Labopette models 9475406, 9475407, 9475503, 9475504, 9475505, 9475506, 9475602, 9475702
€ 70.55
Pack of 50
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
10052173
View Documents Promotion Details
Hirschmann™
9470012
Labopette models 9475408, 9475507, 9475604, 9475704
€ 70.55
Pack of 50
View alternative products
Save Up to 
»
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
10173913
View Documents Promotion Details
Hirschmann™
9470011
Labopette models 9480104, 9480105, 9480106, 9475409, 9475508, 9475509, 9475510, 9480800
€ 70.55
Pack of 50
View alternative products
Save Up to 
»
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Description
Description
  • Provides the decisive advantage when protecting pipettes and samples against contamination
  • Extends the service life of pipettes
  • Provides excellent protection for the internal components if replaced regularly
  • Can serve as additional protection against penetration of liquids into the interior of the pipette
  • For use with a range of Labopette™ models
Specifications
Specifications
Safety filter
Protection against cross-contamination