Learn More
DWK Life Sciences Safeguarding, for safety standard ground sockets
Shop All DWK Life Sciences Products
Click to view available options
Joint Size:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
Unit Size:
Each
Pack of 10
Description
Polyamide tension rings have very good sliding properties. They help to safely loosen fixed joints through even distribution of force.
- By means of an even force distribution, stuck joints can be safely loosened.
- Due to their material properties, pull rings have very good sliding properties.
Specifications
Specifications
| Quantity | 10/Pk. |
| Item Description | To secure the security grinding |
| Type | Safeguard |
| Joint Size | NS 34/35 |