Learn More
Justrite™ Safety Can Type Ii For Flammables & Fuel
Brand:  Justrite™ 7225230Z
Additional Details : Weight : 0.80000kg
Product Code. 11954457
  • € 169.00 / Each
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Description
Description
With hose, SafeSqueeze trigger for maximum pouring control, steel, manifold automatically vents for smooth flow yellow 9.5L Accuflow