Learn More
Pacific Handy Cutter Safety Cutter 3-in-1 (Cutter, Tape Splitter, Film Cutter)
Product Code. 17759037
Change view
Click to view available options
Color:
Green
Red
Handle Type:
Left-Hand
Right-hand
Unit Size:
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Handle Type unitSize
17759037 Red Left-Hand Pack of 12
17769037 Green Right-hand Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17759037 Supplier Pacific Handy Cutter Supplier No. S5L
  • ONLINE EXCLUSIVE PRICE
    € 232.00
    € 198.81 / Pack of 12
    Save € 33.19
    14% Off
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
The 3-in-1 Safety Cutter is a big hit in retail and supermarkets! It has three adjustable blade lengths for box topping, for shallow window cuts or for deep cuts.
  • The extremely stable metal blade guard protects the user.
  • At the same time, the angled guard serves as a cutting guide, e.g. when topping boxes.
  • At the end of the handle there is a fold-out, film cutter with concealed blade, which is ideal for foils and strapping.
  • Integrated robust tape splitter.
  • Spare blades can be comfortably placed under control in the compartment.
  • Tool-free blade change.
  • With practical eyelet for lanyard fastening or hanging.
  • The green model is suitable for right-handed users.
  • The red model is suitable for left-handed users.
Specifications
Product Type Safety Cutter
Handle Type Left-Hand
Disposable Reusable
Sterility Non-sterile
Color Red