Learn More
3M™ Safety Helmets
Product Code. 10628205
Click to view available options
Product Type:
Respirator Helmet
Unit Size:
Each
Product Code. 10628205
Brand: 3M™ 0610082P
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Color White
Product Type Respirator Helmet
For Use With (Equipment) 3M Powered and supplied air respirator systems