Learn More
SCAT Safety Waste Cap 'LISA', V3.0 Blind Plug
Product Code. 17316429 Shop All SCAT Europe Products
Change view
Click to view available options
Type:
Electrically Conductive
Not Electrically Conductive
Thread Size:
GL14
M30x35
NPT 1/8 in.
Includes:
Blind plug
Satellite Blind Plug
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Thread Size Includes unitSize
17316429 Electrically Conductive GL14 Blind plug Each
17326429 Electrically Conductive NPT 1/8 in. Blind plug Each
17336429 Not Electrically Conductive M30x35 Satellite Blind Plug Each
17366429 Electrically Conductive M30x35 Satellite Blind Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17316429 Supplier SCAT Supplier No. 107680
Only null left
Add to basket
Add to basket
Safety Waste Cap 'LISA', V3.0 Blind Plug is an accessory suitable for safely closing unused threaded holes in devices for the safe disposal of liquid HPLC/UHPLC waste.
  • Accessory for Safety Waste Cap 'LISA,' V3.0
  • For extending and attaching various components such as a funnel
  • Made of PTFE for optimal chemical resistance
Specifications
Type Electrically Conductive
Includes Blind plug
Thread Size GL14
For Use With HPLC / UHPLC Systems
Material PTFE