Learn More
SymLine™ Safety Waste Caps
These Safety Waste Caps safely connect the pipework to the waste container as an integrated exhaust filter blocks solvent vapors for health and environmental protection.
Brand: SymLine™ 106484
Additional Details : Weight : 0.20000kg
Description
- Available with different thread sizes for attachment to pipe system
- Different numbers of hose connections for disposal of liquid waste
- Available with electronic level sensors that indicate the degree of filling of the disposal container
- By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
Specifications
Electronic Level Control, GL25 (m), Connector for Exhaust Filter | |
Safety Waste Cap | |
S90 |
Waste Disposal | |
SymLine System |