Learn More
SAG Monoclonal Antibody (OTI7A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA812590
Encompass_Preferred
 View more versions of this product
Product Code. 17597144
  • € 540.87 / 100µL
Description
S-antigen, also identified as arrestin, is a protein involved in the regulation of phototransduction.
Specifications
Specifications
SAG
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P10523
SAG
Full length human recombit protein of human SAG produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI7A12
Unconjugated
SAG
RP47, S-AG
Mouse
Affinity Chromatography
RUO
6295
-20° C, Avoid Freeze/Thaw Cycles