missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAH3 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98616JF646
This item is not returnable.
View return policy
Description
SAH3 Polyclonal antibody specifically detects SAH3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SAH3 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| adenosylhomocysteinase-like 2, AdoHcyase 3, ADOHCYASE3, EC 3.3.1.1, FLJ21719, KIAA0828S-adenosylhomocysteine hydrolase-like 2, putative adenosylhomocysteinase 3, S-adenosylhomocysteine hydrolase-like protein 2, S-adenosyl-L-homocysteine hydrolase 3 | |
| Produced in rabbits immunized with E. coli-derived Human SAH3 fragment. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 23382 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction