Learn More
Saint Gobain Life Sciences Chemware™ PTFE Solid Balls
Click to view available options
Diameter (Metric):
3 mm
6 mm
9.5 mm
Unit Size:
Pack of 100
Description
- For column packing and as mixing beads
- Inert to most liquids and vapors
- Usable to 220°C (428°F)
Specifications
Specifications
| Type | Boiling Stone |
| Material | PTFE |
| Diameter (Metric) | 6 mm |