Learn More
Saint-Gobain Tygon™ Fuel and Lubricant Tubing, Formula F-4040-A
Click to view available options
Diameter (English) Outer:
3/8 in.
Maximum Pressure:
30 psi
Thickness (English):
0.0625 in.
Unit Size:
Each
Description
For transporting gasoline, heating oils, kerosene, and glycol-based coolants.
- Resists swelling, cracking and brittleness
- Compatible with most petroleum-based products
- Ozone and UV light resistant
- Minimum extractability safeguards fluids being handled and equipment fluid contacts
- Not for use with strong acids or alkalis; not to be used in contact with food or beverages
- Distinctive yellow color
Order Info
Durometer hardness: Shore A, 57. In pack of 50 ft. (15m).
Specifications
Specifications
| Diameter (English) Inner | 1/4 in. |
| Material | Tygon |
| Diameter (English) Outer | 3/8 in. |
| Overall Length (English) | 50 ft. |
| Thickness (English) | 0.0625 in. |
| For Use With (Application) | For transporting gasoline, heating oils, kerosene, and glycol-based coolants |
| Min. Operating Temperature (Metric) | -37°C |
| Color | Yellow |
| Maximum Pressure | 30 psi |
| Max. Operating Temperature (Metric) | 74°C |