missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Sal 003
GREENER_CHOICE

Product Code. 18095702
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18095702

Brand: Tocris Bioscience™ 3657/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cell-permeable inhibitor of eIF2α dephosphorylation

Sal 003 is a cell-permeable inhibitor of cellular phosphatase complexes that dephosphorylate eukaryotic translation initiation factor 2 subunit α (eIF2α). Analog of salubrinal (Cat. No. 2347) with improved aqueous solubility. Shown to prevent the induction of hippocampal long-term potentiation (LTP) and memory formation (LTM) in mice.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type Sal 003
Purity >99%
Molecular Weight (g/mol) 463.21
Inhibitors Sal 003

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.